Assay Details
| Gene Symbol: lgi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078898.1 | NP_001072366.1 | 5 - 6 | 762 | 81 | |
| XM_012966735.1 | XP_012822189.1 | 4 - 5 | 817 | 81 | ||
| GenBank mRNA | BC121468.1 | - | 5 - 6 | 762 | 81 | |
More Information
Set Membership: |
|

lgi1
Genomic Map