Assay Details
| Gene Symbol: tdgf1.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078927.1 | NP_001072395.1 | 5 - 6 | 567 | 72 | |
| GenBank mRNA | BC121594.1 | - | 5 - 6 | 567 | 72 | |
| BC157155.1 | - | 5 - 6 | 590 | 72 | ||
| CR760084.2 | - | 5 - 6 | 523 | 72 | ||
More Information
Set Membership: |
|

tdgf1.3
Genomic Map