Assay Details
| Gene Symbol: lactb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078957.1 | NP_001072425.1 | 2 - 3 | 334 | 64 | |
| GenBank mRNA | BC121666.1 | - | 2 - 3 | 334 | 64 | |
More Information
Set Membership: |
|

lactb2
Genomic Map