Assay Details
| Gene Symbol: atg4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078996.1 | NP_001072464.1 | 11 - 12 | 1005 | 62 | |
| XM_012970551.2 | XP_012826005.1 | 11 - 12 | 1062 | 62 | ||
| XM_018093940.1 | XP_017949429.1 | 11 - 12 | 1070 | 62 | ||
| GenBank mRNA | BC121870.1 | - | 11 - 12 | 1005 | 62 | |
More Information
Set Membership: |
|

atg4b
Genomic Map