Assay Details
| Gene Symbol: dedd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079013.1 | NP_001072481.1 | 2 - 3 | 688 | 81 | |
| XM_012966743.1 | XP_012822197.1 | 2 - 3 | 716 | 81 | ||
| XM_018095434.1 | XP_017950923.1 | 3 - 4 | 789 | 81 | ||
| GenBank mRNA | BC121906.1 | - | 2 - 3 | 688 | 81 | |
More Information
Set Membership: |
|

dedd2
Genomic Map