Assay Details
| Gene Symbol: ca6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079014.1 | NP_001072482.1 | 4 - 5 | 511 | 104 | |
| GenBank mRNA | BC121907.1 | - | 4 - 5 | 511 | 104 | |
More Information
Set Membership: |
|

ca6
Genomic Map