Assay Details
| Gene Symbol: isoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079129.1 | NP_001072597.1 | 4 - 5 | 844 | 75 | |
| XM_012965593.2 | XP_012821047.1 | 4 - 5 | 800 | 75 | ||
| XM_012965599.2 | XP_012821053.1 | 4 - 5 | 1339 | 75 | ||
| XM_018094550.1 | XP_017950039.1 | 4 - 5 | 720 | 75 | ||
| GenBank mRNA | BC122966.1 | - | 4 - 5 | 844 | 75 | |
More Information
Set Membership: |
|

isoc1
Genomic Map