Assay Details
| Gene Symbol: otud4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079130.1 | NP_001072598.1 | 9 - 10 | 1174 | 66 | |
| GenBank mRNA | BC122971.1 | - | 9 - 10 | 1174 | 66 | |
More Information
Set Membership: |
|

otud4
Genomic Map