Assay Details
| Gene Symbol: ssbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079190.2 | NP_001072658.1 | 5 - 6 | 411 | 101 | |
| XM_012959268.2 | XP_012814722.1 | 5 - 6 | 451 | 101 | ||
| GenBank mRNA | BC123945.1 | - | 5 - 6 | 375 | 101 | |
| BC135398.1 | - | 5 - 6 | 375 | 101 | ||
| CX469111.2 | - | 2 - 3 | 260 | 101 | ||
More Information
Set Membership: |
|

ssbp1
Genomic Map