Assay Details
| Gene Symbol: ssbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079190.2 | NP_001072658.1 | 6 - 7 | 495 | 73 | |
| XM_012959268.2 | XP_012814722.1 | 6 - 7 | 535 | 73 | ||
| GenBank mRNA | BC123945.1 | - | 6 - 7 | 459 | 73 | |
| BC135398.1 | - | 6 - 7 | 459 | 73 | ||
More Information
Set Membership: |
|

ssbp1
Genomic Map