Assay Details
| Gene Symbol: arg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079217.1 | NP_001072685.1 | 3 - 4 | 387 | 67 | |
| GenBank mRNA | BC124018.1 | - | 3 - 4 | 387 | 67 | |
More Information
Set Membership: |
|

arg2
Genomic Map