Assay Details
| Gene Symbol: map7d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079229.1 | NP_001072697.1 | 16 - 17 | 2360 | 69 | |
| XM_012957301.2 | XP_012812755.1 | 17 - 18 | 2662 | 69 | ||
| XM_012957302.2 | XP_012812756.1 | 17 - 18 | 2640 | 69 | ||
| XM_012957303.2 | XP_012812757.1 | 17 - 18 | 2268 | 69 | ||
| XM_012957304.2 | XP_012812758.1 | 16 - 17 | 2515 | 69 | ||
| XM_012957305.2 | XP_012812759.1 | 15 - 16 | 2409 | 69 | ||
| XM_018091363.1 | XP_017946852.1 | 15 - 16 | 2447 | 69 | ||
| GenBank mRNA | BC124039.1 | - | 16 - 17 | 2360 | 69 | |
More Information
Set Membership: |
|

map7d2
Genomic Map