Assay Details
| Gene Symbol: acp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079268.1 | NP_001072736.1 | 587 | 73 | ||
| GenBank mRNA | BC125717.1 | - | 587 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

acp6| Gene Symbol: acp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079268.1 | NP_001072736.1 | 587 | 73 | ||
| GenBank mRNA | BC125717.1 | - | 587 | 73 | ||
Set Membership: |
|