Assay Details
| Gene Symbol: rnf111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079337.1 | NP_001072805.1 | 7 - 8 | 2089 | 66 | |
| XM_012970566.2 | XP_012826020.1 | 8 - 9 | 2308 | 66 | ||
| XM_012970567.2 | XP_012826021.1 | 8 - 9 | 2181 | 66 | ||
| XM_012970568.1 | XP_012826022.1 | 7 - 8 | 2016 | 66 | ||
| XM_012970569.2 | XP_012826023.1 | 8 - 9 | 2308 | 66 | ||
| XM_012970570.2 | XP_012826024.1 | 8 - 9 | 2308 | 66 | ||
| XM_012970571.2 | XP_012826025.1 | 8 - 9 | 2308 | 66 | ||
| XM_012970572.2 | XP_012826026.1 | 7 - 8 | 2123 | 66 | ||
| GenBank mRNA | BC121428.1 | - | 7 - 8 | 2089 | 66 | |
More Information
Set Membership: |
|

rnf111
Genomic Map