Assay Details
Gene Symbol: kmt5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079347.1 | NP_001072815.1 | 6 - 7 | 752 | 70 | |
XM_018094658.1 | XP_017950147.1 | 5 - 6 | 700 | 70 | ||
GenBank mRNA | BC121601.1 | - | 6 - 7 | 752 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
