Assay Details
| Gene Symbol: fra10ac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079356.1 | NP_001072824.1 | 7 - 8 | 607 | 77 | |
| GenBank mRNA | BC121693.1 | - | 7 - 8 | 607 | 77 | |
More Information
Set Membership: |
|

fra10ac1
Genomic Map