Assay Details
| Gene Symbol: mrpl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079446.1 | NP_001072914.1 | 3 - 4 | 215 | 67 | |
| XM_012959300.2 | XP_012814754.1 | 3 - 4 | 342 | 67 | ||
| XM_012959301.2 | XP_012814755.1 | 3 - 4 | 329 | 67 | ||
| XM_018092187.1 | XP_017947676.1 | 2 - 3 | 217 | 67 | ||
| GenBank mRNA | BC118825.1 | - | 3 - 4 | 215 | 67 | |
More Information
Set Membership: |
|

mrpl22
Genomic Map