Assay Details
| Gene Symbol: pik3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079460.1 | NP_001072928.1 | 21 - 22 | 3026 | 98 | |
| GenBank mRNA | BC122988.1 | - | 21 - 22 | 3026 | 98 | |
More Information
Set Membership: |
|

pik3cb
Genomic Map