Assay Details
| Gene Symbol: smim14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016053.3 | NP_001016053.2 | 3 - 4 | 185 | 65 | |
| GenBank mRNA | BC121508.1 | - | 3 - 4 | 185 | 65 | |
More Information
Set Membership: |
|

smim14
Genomic Map