Assay Details
| Gene Symbol: h1foo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR761180.2 | - | 2 - 3 | 346 | 76 | |
| CX485201.2 | - | 2 - 3 | 344 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

h1foo| Gene Symbol: h1foo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR761180.2 | - | 2 - 3 | 346 | 76 | |
| CX485201.2 | - | 2 - 3 | 344 | 76 | ||
Set Membership: |
|