Assay Details
| Gene Symbol: camlg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016600.4 | NP_001016600.2 | 2 - 3 | 699 | 71 | |
| NM_001244908.1 | NP_001231837.1 | 2 - 3 | 492 | 71 | ||
| GenBank mRNA | BC136163.1 | - | 2 - 3 | 699 | 71 | |
| CR760559.2 | - | 2 - 3 | 607 | 71 | ||
| DR843003.1 | - | 2 - 3 | 492 | 71 | ||
More Information
Set Membership: |
|

camlg
Genomic Map