Assay Details
| Gene Symbol: klhl32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016857.3 | NP_001016857.2 | 8 - 9 | 1563 | 62 | |
| XM_012963031.2 | XP_012818485.1 | 8 - 9 | 2082 | 62 | ||
| XM_012963034.2 | XP_012818488.1 | 7 - 8 | 2009 | 62 | ||
| XM_012963035.2 | XP_012818489.1 | 8 - 9 | 1906 | 62 | ||
| GenBank mRNA | BC136224.1 | - | 8 - 9 | 1563 | 62 | |
| CR855406.2 | - | 7 - 8 | 1595 | 62 | ||
More Information
Set Membership: |
|

klhl32
Genomic Map