Assay Details
| Gene Symbol: aurkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016859.3 | NP_001016859.2 | 8 - 9 | 952 | 71 | |
| XM_012953827.2 | XP_012809281.1 | 7 - 8 | 963 | 71 | ||
| XM_012953828.2 | XP_012809282.1 | 8 - 9 | 998 | 71 | ||
| GenBank mRNA | BC135173.1 | - | 8 - 9 | 952 | 71 | |
| CT027921.1 | - | 7 - 8 | 913 | 71 | ||
More Information
Set Membership: |
|

aurkb
Genomic Map