Assay Details
| Gene Symbol: gtf2e1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079088.1 | NP_001072556.1 | 2 - 3 | 799 | 77 | |
| XM_012965480.2 | XP_012820934.1 | 3 - 4 | 1691 | 77 | ||
| XM_018094504.1 | XP_017949993.1 | 3 - 4 | 1813 | 77 | ||
| GenBank mRNA | BC122072.1 | - | 2 - 3 | 799 | 77 | |
More Information
Set Membership: |
|

gtf2e1.2
Genomic Map