Assay Details
| Gene Symbol: napb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079098.1 | NP_001072566.1 | 10 - 11 | 821 | 74 | |
| XM_012963158.2 | XP_012818612.1 | 9 - 10 | 1096 | 74 | ||
| XM_018093944.1 | XP_017949433.1 | 10 - 11 | 781 | 74 | ||
| GenBank mRNA | BC122906.1 | - | 10 - 11 | 821 | 74 | |
| CT030511.1 | - | 10 - 11 | 827 | 74 | ||
More Information
Set Membership: |
|

napb
Genomic Map