Assay Details
| Gene Symbol: tbc1d12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079100.1 | NP_001072568.1 | 10 - 11 | 1778 | 57 | |
| GenBank mRNA | BC122911.1 | - | 10 - 11 | 1778 | 57 | |
More Information
Set Membership: |
|

tbc1d12
Genomic Map