Assay Details
| Gene Symbol: usp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079113.1 | NP_001072581.1 | 5 - 6 | 771 | 75 | |
| XM_012961351.2 | XP_012816805.1 | 5 - 6 | 1059 | 75 | ||
| XM_012961354.2 | XP_012816808.1 | 5 - 6 | 1061 | 75 | ||
| GenBank mRNA | BC122939.1 | - | 5 - 6 | 771 | 75 | |
| CR762284.2 | - | 5 - 6 | 709 | 75 | ||
| CU024926.1 | - | 5 - 6 | 834 | 75 | ||
More Information
Set Membership: |
|

usp1
Genomic Map