Assay Details
| Gene Symbol: galnt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079237.2 | NP_001072705.1 | 10 - 11 | 1633 | 102 | |
| GenBank mRNA | BC124051.1 | - | 9 - 10 | 1550 | 102 | |
| BC135911.1 | - | 10 - 11 | 1633 | 102 | ||
More Information
Set Membership: |
|

galnt4
Genomic Map