Assay Details
| Gene Symbol: rtn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085350.2 | NP_001078819.2 | 5 - 6 | 2135 | 69 | |
| XM_012968532.1 | XP_012823986.1 | 3 - 4 | 606 | 69 | ||
| GenBank mRNA | BC118846.1 | - | 3 - 4 | 522 | 69 | |
| BC152060.1 | - | 5 - 6 | 2135 | 69 | ||
| BC154675.1 | - | 3 - 4 | 521 | 69 | ||
More Information
Set Membership: |
|

rtn1
Genomic Map