Assay Details
| Gene Symbol: dhh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097169.1 | NP_001090638.1 | 2 - 3 | 1144 | 75 | |
| GenBank mRNA | BC125725.1 | - | 2 - 3 | 1144 | 75 | |
More Information
Set Membership: |
|

dhh
Genomic Map