Assay Details
| Gene Symbol: neurl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097237.1 | NP_001090706.1 | 1 - 2 | 134 | 64 | |
| GenBank mRNA | BC128632.1 | - | 1 - 2 | 134 | 64 | |
More Information
Set Membership: |
|

neurl1
Genomic Map