Assay Details
| Gene Symbol: pou3f4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097259.1 | NP_001090728.1 | 1 | 1047 | 103 | |
| GenBank mRNA | BC127328.1 | - | 1 | 1047 | 103 | |
More Information
Set Membership: |
|

pou3f4
Genomic Map