Assay Details
| Gene Symbol: lamp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097312.1 | NP_001090781.1 | 4 - 5 | 760 | 76 | |
| XM_012962533.2 | XP_012817987.1 | 5 - 6 | 1361 | 76 | ||
| GenBank mRNA | BC135151.1 | - | 4 - 5 | 760 | 76 | |
More Information
Set Membership: |
|

lamp5
Genomic Map