Assay Details
| Gene Symbol: cyp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097344.1 | NP_001090813.1 | 6 - 7 | 1372 | 67 | |
| GenBank mRNA | BC135260.1 | - | 6 - 7 | 1372 | 67 | |
| HQ018040.1 | - | 6 - 7 | 1306 | 67 | ||
More Information
Set Membership: |
|

cyp1a1
Genomic Map