Assay Details
| Gene Symbol: pitpna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097363.1 | NP_001090832.1 | 11 - 12 | 875 | 84 | |
| XM_018091036.1 | XP_017946525.1 | 11 - 12 | 1031 | 84 | ||
| GenBank mRNA | BC135421.1 | - | 11 - 12 | 875 | 84 | |
| CT030292.1 | - | 11 - 12 | 876 | 84 | ||
More Information
Set Membership: |
|

pitpna
Genomic Map