Assay Details
| Gene Symbol: wwp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097372.1 | NP_001090841.1 | 9 - 10 | 852 | 73 | |
| XM_012960385.2 | XP_012815839.1 | 8 - 9 | 1636 | 73 | ||
| XM_012960386.2 | XP_012815840.1 | 2 - 3 | 380 | 73 | ||
| GenBank mRNA | BC121316.1 | - | 9 - 10 | 852 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_012960386.2 transcripts.
Set Membership: |
|

wwp2
Genomic Map