Assay Details
| Gene Symbol: mrps6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097389.1 | NP_001090858.1 | 2 - 3 | 266 | 91 | |
| GenBank mRNA | BC135212.1 | - | 2 - 3 | 266 | 91 | |
| CT030115.2 | - | 2 - 3 | 351 | 91 | ||
More Information
Set Membership: |
|

mrps6
Genomic Map