Assay Details
| Gene Symbol: slc39a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097395.1 | NP_001090864.1 | 7 - 8 | 1249 | 70 | |
| XM_012958370.2 | XP_012813824.1 | 7 - 8 | 1361 | 70 | ||
| XM_012958371.2 | XP_012813825.1 | 7 - 8 | 1364 | 70 | ||
| XM_012958372.2 | XP_012813826.1 | 7 - 8 | 1288 | 70 | ||
| GenBank mRNA | BC135298.1 | - | 7 - 8 | 1249 | 70 | |
More Information
Set Membership: |
|

slc39a14
Genomic Map