Assay Details
| Gene Symbol: mydgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100209.1 | NP_001093679.1 | 2 - 3 | 231 | 96 | |
| GenBank mRNA | BC135669.1 | - | 2 - 3 | 231 | 96 | |
More Information
Set Membership: |
|

mydgf
Genomic Map