Assay Details
| Gene Symbol: ins | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100236.1 | NP_001093706.1 | 2 - 3 | 229 | 72 | |
| GenBank mRNA | BC135265.1 | - | 2 - 3 | 229 | 72 | |
More Information
Set Membership: |
|

ins
Genomic Map