Assay Details
| Gene Symbol: efnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100239.1 | NP_001093709.1 | 3 - 4 | 969 | 93 | |
| GenBank mRNA | BC135636.1 | - | 3 - 4 | 969 | 93 | |
More Information
Set Membership: |
|

efnb2
Genomic Map