Assay Details
| Gene Symbol: smn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100240.1 | NP_001093710.1 | 2 - 3 | 177 | 88 | |
| GenBank mRNA | BC135221.1 | - | 2 - 3 | 177 | 88 | |
More Information
Set Membership: |
|

smn2
Genomic Map