Assay Details
| Gene Symbol: smn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100240.1 | NP_001093710.1 | 6 - 7 | 726 | 68 | |
| XM_018092087.1 | XP_017947576.1 | 5 - 6 | 619 | 68 | ||
| GenBank mRNA | BC135221.1 | - | 6 - 7 | 726 | 68 | |
More Information
Set Membership: |
|

smn2
Genomic Map