Assay Details
| Gene Symbol: kazald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100263.1 | NP_001093733.1 | 4 - 5 | 1049 | 63 | |
| GenBank mRNA | BC135810.1 | - | 4 - 5 | 1049 | 63 | |
More Information
Set Membership: |
|

kazald1
Genomic Map