Assay Details
| Gene Symbol: mitf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100277.1 | NP_001093747.1 | 1208 | 56 | ||
| GenBank mRNA | BC135968.1 | - | 1208 | 56 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mitf| Gene Symbol: mitf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100277.1 | NP_001093747.1 | 1208 | 56 | ||
| GenBank mRNA | BC135968.1 | - | 1208 | 56 | ||
Set Membership: |
|