Assay Details
| Gene Symbol: otub2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101812.1 | NP_001095282.1 | 4 - 5 | 307 | 77 | |
| XM_012967849.2 | XP_012823303.2 | 11 - 12 | 3471 | 77 | ||
| GenBank mRNA | BC135508.1 | - | 4 - 5 | 307 | 77 | |
More Information
Set Membership: |
|

otub2
Genomic Map