Assay Details
| Gene Symbol: rcan3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102693.1 | NP_001096163.1 | 5 - 6 | 802 | 70 | |
| GenBank mRNA | BC146616.1 | - | 5 - 6 | 802 | 70 | |
More Information
Set Membership: |
|

rcan3
Genomic Map