Assay Details
| Gene Symbol: aip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102749.1 | NP_001096219.1 | 3 - 4 | 481 | 85 | |
| XM_012965878.2 | XP_012821332.1 | 3 - 4 | 586 | 85 | ||
| GenBank mRNA | BC158230.1 | - | 3 - 4 | 479 | 85 | |
| CR762165.2 | - | 3 - 4 | 481 | 85 | ||
More Information
Set Membership: |
|

aip
Genomic Map