Assay Details
Gene Symbol: usf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102766.1 | NP_001096236.1 | 422 | 61 | ||
GenBank mRNA | BC135362.1 | - | 422 | 61 | ||
CT030570.1 | - | 356 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
