Assay Details
| Gene Symbol: mtfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102827.1 | NP_001096297.1 | 2 - 3 | 281 | 61 | |
| GenBank mRNA | BC135607.1 | - | 2 - 3 | 281 | 61 | |
More Information
Set Membership: |
|

mtfp1
Genomic Map